missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium hydrogen phosphate dihydrate, 98% min
Description
Calcium hydrogen phosphate dihydrate is used in the manufacture of pharmaceutical tablets and as a source of calcium and phosphorus in nutritional supplements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 109°C (in -H2O) |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 kg |
| Odor | Odorless |
| Density | 2.306 g/mL |
| Linear Formula | CaHPO4·2H2O |
| Merck Index | 14,1692 |
| Solubility Information | Slightly soluble in water; Soluble in dilute hydrochloric, nitric, and acetic acid; Insoluble in alcohol |
| Formula Weight | 172.09 (136.06 Anhydrous) |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 231-826-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only