missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydrogen phosphate, anhydrous, 98% min

Catalog No. p-7051003
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4023230 250 g
AA40232A1 1 kg
AA40232A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4023230 Supplier Thermo Scientific Chemicals Supplier No. 04023230
Only null left

CAS: 7757-93-9 | CaHO4P | 136.06 g/mol

It is used as a feed additive for livestock and poultry. It can promote feed digestion, put on animal’s weight, raise laying rate or producing milk rate, cure diseases such as rickets, osteomalacia and anemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
PubChem CID 24441
ChEBI CHEBI:32596
IUPAC Name calcium;hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 250 g
Linear Formula CaHPO4
Merck Index 14,1692
Solubility Information Soluble in hydrochloric acid,nitric acid,acetic acid,ammonium citrate solution. Slightly soluble in water. Insoluble in ethanol.
Formula Weight 136.06
Percent Purity ≤98%
Odor Odorless
Chemical Name or Material Calcium hydrogen phosphate, Anhydrous
EINECSNumber 231-826-1
RTECSNumber TB8528000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only