missing translation for 'onlineSavingsMsg'
Learn More

Calcium Gluconate Monohydrate 98.0+%, TCI America™

Catalog No. G003725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G003725G 25 g
G0037500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G003725G Supplier TCI America Supplier No. G003725G

Chemical Identifiers

CAS 299-28-5
Molecular Formula C12H22CaO14
Molecular Weight (g/mol) 430.372
MDL Number MFCD00064209
InChI Key NEEHYRZPVYRGPP-IYEMJOQQSA-L
Synonym calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal
PubChem CID 9290
IUPAC Name calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]

Specifications

Color White
Quantity 25 g
Formula Weight 430.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Gluconate Monohydrate
EINECSNumber (2)-1410
RTECSNumber EW2100000
TSCA Yes