Learn More
Calgonate CALCIUM GLUCONATE 2.5% GEL 25G
Supplier:  Calgonate 06306
Encompass
Per supplier requirements this item has a minimum qty 3. Calgonate Gel is a topical 2.5% calcium gluconate gel that is used in first aid response to HF acid exposure to the body. Calgonate contains Calcium Gluconate USP 2.5%. It is manufactured in Rhode Island at an FDA-GMP approved facility
Catalog No. NC9966227