Learn More
Calcium Formate, 98%, Pure
CAS: 544-17-2 | C2H2CaO4 | 130.11 g/mol
Supplier: Thermo Scientific Chemicals 403830010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Calcium formate | |
| >300.0°C | |
| Authentic | |
| Glass bottle | |
| (HCOO)2Ca | |
| 1 kg | |
| calcium formate, calcium diformate, formic acid, calcium salt, calcoform, unii-np3jd65npy, mravencan vapenaty czech, calcium formate ca hco2 2, formic acid calcium salt, np3jd65npy, formic acid, calcium salt 2:1 | |
| CBOCVOKPQGJKKJ-UHFFFAOYSA-L | |
| calcium;diformate | |
| 10997 | |
| 130.11 | |
| Pure |
| 544-17-2 | |
| White | |
| 98% | |
| C2H2CaO4 | |
| MFCD00036108 | |
| 15,167 | |
| Solubility in water: soluble. Other solubilities: insoluble in alcohol | |
| C(=O)[O-].C(=O)[O-].[Ca+2] | |
| 130.11 | |
| CHEBI:81851 | |
| 98% | |
| Fine Crystalline Powder |
Chemical Identifiers
| 544-17-2 | |
| 130.11 | |
| CBOCVOKPQGJKKJ-UHFFFAOYSA-L | |
| 10997 | |
| calcium;diformate |
| C2H2CaO4 | |
| MFCD00036108 | |
| calcium formate, calcium diformate, formic acid, calcium salt, calcoform, unii-np3jd65npy, mravencan vapenaty czech, calcium formate ca hco2 2, formic acid calcium salt, np3jd65npy, formic acid, calcium salt 2:1 | |
| CHEBI:81851 | |
| C(=O)[O-].C(=O)[O-].[Ca+2] |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 208-863-7
RUO – Research Use Only