missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium fluoride, Reagent Grade, 97%
Description
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers, electronics, lasers, high-temperature dry-film lubricants, emery wheels, certain cements, phosphors, paint pigment, catalyst in wood preservatives, and optical equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | ca 2500°C |
| Melting Point | 1403°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 97% |
| Solubility Information | Insoluble in water. Soluble in ammonium salts. Soluble in conc. mineral acids with evolution of HF |
| IUPAC Name | calcium;difluoride |
| Formula Weight | 78.08 |
| Grade | Reagent |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335-H303 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed. |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. |
| EINECSNumber | 232-188-7 |
| RTECSNumber | EW1760000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only