missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, Puratronic™, 99.985% (metals basis)

Catalog No. AA1068104
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1068104 2 g
AA1068109 10 g
AA1068118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1068104 Supplier Thermo Scientific Chemicals Supplier No. 1068104

CAS: 7789-75-5 | CaF2 | 78.08 g/mol

Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
SMILES [F-].[F-].[Ca++]

Specifications

Boiling Point ∼2500°C
Melting Point 1403°C
Color White
Physical Form Powder
Quantity 2 g
Merck Index 14,1667
Solubility Information Soluble in water (slightly).
IUPAC Name calcium difluoride
Formula Weight 78.08
Percent Purity 99.985%
Grade Puratronic
Odor Odorless
Refractive Index 1.434
Sensitivity Hygroscopic
Density 3.18 g/mL
Assay (metals basis)
Chemical Name or Material Calcium fluoride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 232-188-7
RTECSNumber EW1760000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only