missing translation for 'onlineSavingsMsg'
Learn More

Calcium Fluoride, Powder, 97%, Spectrum™ Chemical

Catalog No. 18609477
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609477 125 g Poly Bottle
18609479 500 g Poly Bottle
18609481 2.5 kg Poly Pail
18609478 12 kg Poly Pail
18609480 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609477 Supplier Spectrum Chemical Mfg Cor Supplier No. C1105125GM

Chemical Identifiers

CAS 7789-75-5

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 97%
Grade Reagent
Loss on Drying 0.01
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.