missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, 99%

Catalog No. AAA1091336
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAA1091336 500 g Plastic bottle
AAA1091322 100 g Glass bottle
AAA109130E 2500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1091336 Supplier Thermo Scientific Chemicals Supplier No. A1091336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Commercially important source for fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. | CAS: 7789-75-5 | CaF2 | 78.08 g/mol

Calcium fluoride is the commercially important source for fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers; it is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
SMILES [F-].[F-].[Ca++]

Specifications

Boiling Point ≈2500°C
Melting Point 1403°C
Physical Form Crystals
Quantity 500 g
Beilstein EM>5,
Solubility Information Slightly soluble in acid. Insoluble in acetone.
IUPAC Name calcium difluoride
Formula Weight 78.08
Percent Purity 99%
Packaging Plastic bottle
Refractive Index 1.434
Specific Gravity 3.18
Density 3.18
Chemical Name or Material Calcium fluoride
Show More Show Less
EINECSNumber 232-188-7
RTECSNumber EW1760000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only