missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, 99%, pure, anhydrous

Catalog No. p-222755
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218250100 10 g Plastic bottle
AC218251000 100 g Plastic bottle
AC218255000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC218250100 Supplier Thermo Scientific Chemicals Supplier No. 218250100
Only null left

CAS: 7789-75-5 | CaF2 | 78.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
SMILES [F-].[F-].[Ca++]

Specifications

Boiling Point 2500.0°C
Melting Point 1402.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 98.5% min. (CaF2)
Linear Formula CaF2
Fieser 15,287
Merck Index 15,1669
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonium salts
IUPAC Name calcium;difluoride
Formula Weight 78.08
Percent Purity 99%
Grade Pure
Packaging Plastic bottle
Specific Gravity 3.18
Density 3.1800g/mL
Chemical Name or Material Calcium fluoride, 99%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-188-7
RTECSNumber EW1760000
TSCA TSCA

RUO – Research Use Only