missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, 99.5% (metals basis)

CAS: 7789-75-5 | CaF2 | 78.08 g/mol

$2321.37 - $2321.37

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
SMILES [F-].[F-].[Ca++]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA11055A7
View Documents
Thermo Scientific Chemicals
011055A7
5 kg
Each for $2,321.37
Only null left
 
Description

Description

Calcium fluoride is the commercially important source of fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers. It is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-75-5
78.08
WUKWITHWXAAZEY-UHFFFAOYSA-L
84512
[F-].[F-].[Ca++]
CaF2
MFCD00010907
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
CHEBI:35437

Specifications

ca 2500°C
7789-75-5
Powder
99.5% (Metal basis)
MFCD00010907
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
WUKWITHWXAAZEY-UHFFFAOYSA-L
calcium difluoride
84512
78.08
1.434
White powder
Calcium fluoride
1403°C
White
5 kg
CaF2
EM>5,
Slightly soluble in acid. Insoluble in acetone.
[F-].[F-].[Ca++]
78.08
CHEBI:35437
Odorless
Hygroscopic
3.18
Videos
Safety and Handling

Safety and Handling

H315-H319-H335-H303

EINECSNumber : 232-188-7

RTECSNumber : EW1760000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only