missing translation for 'onlineSavingsMsg'
Learn More

Calcium D-saccharate tetrahydrate, 98%

Catalog No. AAJ6694714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6694714 25 g
AAJ6694722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6694714 Supplier Thermo Scientific Chemicals Supplier No. J6694714
Only null left

CAS: 5793-89-5 | C6H16CaO12 | 320.26 g/mol

D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.

Solubility
Soluble in water(0.43g/L).

Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 5793-89-5
Molecular Formula C6H16CaO12
Molecular Weight (g/mol) 320.26
MDL Number MFCD00150704
InChI Key ZQWFSIZRQANUDA-WQMSYZFBSA-L
Synonym calcium saccharate, unii-6ap9j91k4v, d-saccharic acid calcium salt tetrahydrate, calcium d-glucarate tetrahydrate, calcium d-saccharate tetrahydrate, calcium saccharat-4-wasser, calcium saccharate usan:usp:inn, calcium saccharate usp, calcium d-glucarate, tetrahydrate, calcium d-glucarate 1:1 , tetrahydrate
PubChem CID 16211545
IUPAC Name calcium;(2R,3S,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate;tetrahydrate
SMILES C(C(C(C(=O)[O-])O)O)(C(C(=O)[O-])O)O.O.O.O.O.[Ca+2]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Beilstein 3821334
Merck Index 14,1700
Solubility Information Soluble in water(0.43g/L).
Formula Weight 320.26
Percent Purity 98%
Odor Odorless
Chemical Name or Material Calcium D-saccharate tetrahydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-334-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only