missing translation for 'onlineSavingsMsg'
Learn More

Calcium D-gluconate monohydrate, 98+%

Catalog No. AAA1164922
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1164922 100 g
AAA1164936 500 g
AAA116490E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1164922 Supplier Thermo Scientific Chemicals Supplier No. A1164922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 66905-23-5 | C12H24CaO15 | 448.387 g/mol

Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.

Solubility
Soluble in water(30g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 66905-23-5
MDL Number MFCD00064209

Specifications

Melting Point ∽195°C (decomposition)
Assay Percent Range ≥98%
Molecular Formula C12H22CaO14
Quantity 100 g
Merck Index 14,1670
Synonym calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal
Solubility Information Soluble in water(30g/L).
InChI Key NEEHYRZPVYRGPP-IYEMJOQQSA-L
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
IUPAC Name calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
Molecular Weight (g/mol) 430.372
PubChem CID 9290
Formula Weight 448.40 (430.37 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Calcium D-gluconate Monohydrate
Show More Show Less
EINECSNumber 206-075-8
RTECSNumber EW2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only