missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Hexahydrate (Crystalline/Certified ACS ), Fisher Chemical

Catalog No. C78500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C78500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C78500 Supplier Thermo Fisher Scientific Supplier No. FLC78500
Only null left

Chemical Identifiers

CAS 7774-34-7
Molecular Formula CaCl2H12O6
Molecular Weight (g/mol) 219.068
MDL Number MFCD00149616
InChI Key QHFQAJHNDKBRBO-UHFFFAOYSA-L
Synonym calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g
PubChem CID 6093252
IUPAC Name calcium;dichloride;hexahydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]

Specifications

Melting Point 30°C
Color Colorless
pH 6.5 to 8.0
Physical Form Solid
Quantity 500 g
Assay Percent Range 99 to 103 %
Formula Weight 219.08
Percent Purity 99 to 103%
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Density 1.71g/cm³
Chemical Name or Material Calcium Chloride Hexahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage Refrigerate