missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride hexahydrate, 97+%, extra pure

Catalog No. p-3527063
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC389250010 1 kg Plastic bottle
AC389250025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC389250010 Supplier Thermo Scientific Chemicals Supplier No. 389250010
Only null left

CAS: 7774-34-7 | CaCl2·6H2O | 219.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7774-34-7
Molecular Formula CaCl2·6H2O
Molecular Weight (g/mol) 219.08
MDL Number MFCD00149616
InChI Key QHFQAJHNDKBRBO-UHFFFAOYSA-L
Synonym calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g
PubChem CID 6093252
IUPAC Name calcium;dichloride;hexahydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]

Specifications

Melting Point 30.0°C
Color Colorless
Physical Form Chunks
Quantity 1 kg
Assay Percent Range 97.0 to 103% (Complexometry)
Linear Formula CaCl2·6H2O
Merck Index 15, 1661
Solubility Information Solubility in water: 5360g/L (20°C)
Formula Weight 219.08
Percent Purity ≥97%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Calcium chloride hexahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only