missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Dihydrate (USP/FCC/EP), Fisher Chemical

Catalog No. C69500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C69500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C69500 Supplier Thermo Fisher Scientific Supplier No. C69500
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10035-04-8
Molecular Formula Ca Cl2 · 2 H2O
MDL Number MFCD00149613

Specifications

Melting Point 175°C
Color Beige-White
pH 4.5 to 6.5
Physical Form Solid
Quantity 500 g
Formula Weight 147.01
Percent Purity 97.0 to 103.0% (EP)
Grade EP/FCC/USP
Identification Pass Test (EP), Pass Test (EP/FCC/USP), Pass Test (FCC), Pass Test (USP)
Packaging Poly Bottle
Density 0.830g/cm³
Chemical Name or Material Calcium Chloride Dihydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1