missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Catalog No. p-7072022
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 2.5 kg
Catalog No. Quantity
6002345 1 kg
6002346 2.5 kg
60045942 6 x 2.5 kg
3 options

Catalog No. 6002345

Supplier: Honeywell Chemicals 313071KG

Only null left

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2 · 2H2O

Specifications

Quantity 1 kg
MDL Number MFCD00149613
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Calcium Chloride Dihydrate, Reag. Ph. Eur.
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 600-075-5
RTECSNumber EV9810000