missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride dihydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 6 x 2.5 kg |
| MDL Number | MFCD00149613 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 147.01 |
| Percent Purity | ≥99% |
| Grade | ACS/puriss. p.a. |
| Chemical Name or Material | Calcium Chloride Dihydrate, Reag. Ph. Eur. |
Safety and Handling
| Hazard Statement | P305 + P351 + P338 |
| Precautionary Statement | H319 |
| EINECSNumber | 600-075-5 |
| RTECSNumber | EV9810000 |