missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate, puriss., Honeywell Fluka™
Puriss., meets analytical specification of FCC, Ph. Eur., USP, E509, 99-103%, ≤0.0001% Al
Supplier: Honeywell Chemicals 120226X1KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Calcium Chloride Dihydrate | |
| 10035-04-8 | |
| CaCl2 · 2H2O | |
| NONH for all modes of transport | |
| 99 to 103% |
| Meets analytical specification of Ph. Eur.USP, FCC | |
| 6 x 1 kg | |
| MFCD00149613 | |
| 147.01 | |
| puriss. |
Chemical Identifiers
| 10035-04-8 | |
| CaCl2 · 2H2O | |
| MFCD00149613 |
Safety and Handling
P305 + P351 + P338
H319
EINECSNumber : 233-140-8
RTECSNumber : EV9810000