missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Dihydrate, puriss., Honeywell Fluka™

Catalog No. 6001917
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Catalog No. Quantity
6001917 1 kg
6001918 2.5 kg
60044711 6 x 1 kg
60044712 6 x 2.5 kg
4 options

Catalog No. 6001917

Supplier: Honeywell Chemicals 120221KG

Only null left

Puriss., meets analytical specification of FCC, Ph. Eur., USP, E509, 99-103%, ≤0.0001% Al

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2 · 2H2O
MDL Number MFCD00149613

Specifications

Quantity 1 kg
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity 99 to 103%
Grade puriss.
Chemical Name or Material Calcium Chloride Dihydrate, Meets analytical specification of Ph. Eur.USP, FCC
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 233-140-8
RTECSNumber EV9810000