missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, Dihydrate, MP Biomedicals™

Calcium chloride, Dihydrate Cell Culture Reagent, ∽99%, White Powder

$58.73 - $725.61

Chemical Identifiers

CAS 10035-04-8
MDL Number MFCD00149613
Synonym Calcium dichloride dihydrate
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19481580
View Documents
MP Biomedicals Inc
0219481580
100 g
Each for $58.73
Only null left
 
ICN19481590
View Documents
MP Biomedicals Inc
0219481590
500 g
Each for $107.18
Only null left
 
ICN19463501
View Documents
MP Biomedicals Inc
0219463501
1 kg
Each for $182.87
Only null left
 
ICN19463505
View Documents
MP Biomedicals Inc
0219463505
5 kg
Each for $725.61
Only null left
 
Description

Description

  • Calcium chloride is a commonly used reagent in biochemistry.
  • It plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability.
  • Calcium chloride is used in the preparation and transformation of competent E. coli and in the transfection of eukaryotic cells with either plasmid DNA or high molecular weight genomic DNA. has been used in the stabilization and two dimensional crystallization of the NADH-ubiquinone oxidoreductase (complex I) from Escherichia coli.
  • It is also used as a dessicant, coagulant in rubber manufacturing and as an accelerator in the concrete industry.
  • Calcium Chloride is a salt of calcium and chloride.
  • It behaves as a typical ionic halide, and is solid at room temperature
Specifications

Chemical Identifiers

10035-04-8
MFCD00149613
Calcium dichloride dihydrate

Specifications

176°C (Decomposition)(Literature)
4 to 9.5 (5% aq soln)
100 g
CaCl2·2H2O
Calcium dichloride dihydrate
147
∽99%
10035-04-8
Powder
∽99%
MFCD00149613
Soluble in water - 294mg/mL (clear, colorless solution) with heat liberation, ethanol, acetic acid, acetone.
147
Calcium Chloride Dihydrate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : EV9810000

storageNote1 : Store at Room Temperature (15-30°C). Store Desiccated.

Recommended Storage : Room Temperature

SDS
Documents

Documents