missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, Dihydrate, MP Biomedicals™

Catalog No. ICN19481590
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19481590 500 g
ICN19481580 100 g
ICN19463501 1 kg
ICN19463505 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19481590 Supplier MP Biomedicals Inc Supplier No. 0219481590

Calcium chloride, Dihydrate Cell Culture Reagent, ∽99%, White Powder

  • Calcium chloride is a commonly used reagent in biochemistry.
  • It plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability.
  • Calcium chloride is used in the preparation and transformation of competent E. coli and in the transfection of eukaryotic cells with either plasmid DNA or high molecular weight genomic DNA. has been used in the stabilization and two dimensional crystallization of the NADH-ubiquinone oxidoreductase (complex I) from Escherichia coli.
  • It is also used as a dessicant, coagulant in rubber manufacturing and as an accelerator in the concrete industry.
  • Calcium Chloride is a salt of calcium and chloride.
  • It behaves as a typical ionic halide, and is solid at room temperature

Chemical Identifiers

CAS 10035-04-8
MDL Number MFCD00149613
Synonym Calcium dichloride dihydrate

Specifications

Melting Point 176°C (Decomposition)(Literature)
pH 4 to 9.5 (5% aq soln)
Physical Form Powder
Quantity 500 g
Assay Percent Range ∽99%
Molecular Formula CaCl2·2H2O
Solubility Information Soluble in water - 294mg/mL (clear, colorless solution) with heat liberation, ethanol, acetic acid, acetone.
Molecular Weight (g/mol) 147
Formula Weight 147
Chemical Name or Material Calcium Chloride Dihydrate
Grade ∽99%
Show More Show Less
RTECSNumber EV9810000
storageNote1 Store at Room Temperature (15-30°C). Store Desiccated.
Recommended Storage Room Temperature