missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calcium Chloride Dihydrate, Molecular biology grade, Calbiochem™,

Catalog No. 208291250GM
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
208291250GM 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 208291250GM Supplier MilliporeSigma™ Supplier No. 208291250GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recommended Storage

15°C to 30°C, Stock solutions are Stable for up to 6 months at 20°C

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2 · 2H2O
Synonym CaCl2

Specifications

Color White
Physical Form Solid
Quantity 250 g
Percent Purity ≥99% (complexometric)
Packaging Glass Bottle
Recommended Storage 15°C to 30°C, Stock solutions are Stable for up to 6 months at 20°C