missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, Dihydrate, BiotechGrade, 99-103%, Spectrum™ Chemical
C1086, 10035-04-8, CaCl2·2H2O
$520.47 - $7,970.73
Chemical Identifiers
| CAS | 10035-04-8 |
|---|---|
| Molecular Formula | CaCl2H4O2 |
| Molecular Weight (g/mol) | 147.01 |
| InChI Key | LLSDKQJKOVVTOJ-UHFFFAOYSA-L |
| IUPAC Name | calcium dihydrate dichloride |
| SMILES | O.O.[Cl-].[Cl-].[Ca++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601258
|
Spectrum Chemical Mfg Cor
C1086500GM |
500 g | Poly Bottle |
N/A
|
|
||||
|
18601259
|
Spectrum Chemical Mfg Cor
C108625KG |
2.5 kg | Poly Bottle |
N/A
|
|
||||
|
18601260
|
Spectrum Chemical Mfg Cor
C108612KGBL |
12 kg | Poly Pail |
N/A
|
|
||||
Description
Spectrum™ Chemical Calcium Chloride, Dihydrate, BiotechGrade is a white deliquescent compound which is commonly used as a reagent in biochemistry. Biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability rely on calcium ions. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.Chemical Identifiers
| 10035-04-8 | |
| 147.01 | |
| calcium dihydrate dichloride |
| CaCl2H4O2 | |
| LLSDKQJKOVVTOJ-UHFFFAOYSA-L | |
| O.O.[Cl-].[Cl-].[Ca++] |
Specifications
| 10035-04-8 | |
| 4.5- 9.2 | |
| CaCl2H4O2 | |
| O.O.[Cl-].[Cl-].[Ca++] | |
| 147.01 | |
| Biotech | |
| White crystals |
| 100% | |
| 500 g | |
| LLSDKQJKOVVTOJ-UHFFFAOYSA-L | |
| calcium dihydrate dichloride | |
| 99 to 103% | |
| Poly Bottle |