missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Dihydrate, ACS Reagent, Honeywell Fluka™

Catalog No. p-7071908
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
Catalog No. Quantity
6002149 2.5 kg
6002153 6 x 500 g
2 options

Catalog No. 6002149

Supplier: Honeywell Chemicals 2235062.5KG

Only null left

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2 · 2H2O
MDL Number MFCD00149613

Specifications

Quantity 2.5 kg
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity ≥99%
Grade ACS Reagent
Vapor Pressure 0.01mmHg (20°C)
Chemical Name or Material Calcium Chloride Dihydrate
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 233-140-8
RTECSNumber EV9810000