missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, ACS, 99.0-105.0%

Catalog No. AA33296A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33296A3 2 kg
AA3329636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33296A3 Supplier Thermo Scientific Chemicals Supplier No. 033296A3

CAS: 10035-04-8 | CaCl2·2H2O

Calcium chloride dihydrate is widely used as an ingredient in ceramic slipware, cosmetic and food items. It acts as a pH buffer in swimming pool water. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. It is employed in the oil industry to increase the density of solids-free brines. It is used as a flux in the Davy process for the manufacture of sodium through the electrolysis of molten sodium chloride. It finds an important application as an additive in plastics, fire extinguishers, blast furnaces and in wastewater treatment as a drainage aid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O
MDL Number MFCD00149613

Specifications

Physical Form Powder
Quantity 2 kg
Assay Percent Range 99.0-105.0%
Solubility Information Soluble in water,acetic acid and alcohol.
Formula Weight 147.02 (110.99 Anhydrous)
Grade ACS Reagent
Sensitivity Hygroscopic
Density 0.835 g/mL
Chemical Name or Material Calcium chloride dihydrate, For ACS analysis
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 233-140-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only