missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, Dihydrate, ACS, 99.0 to 102.0%, MilliporeSigma™

Catalog No. M1023825000
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1023825000 5 kg Plastic Bottle
M1023820500 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1023825000 Supplier Technidata Supplier No. 1.02382.5000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·H2O
Synonym Calcium dichloride

Specifications

Color White
Physical Form Powder
Quantity 5 kg
Assay Percent Range 99.0 to 102.0%
Linear Formula CaCl2·H2O
Formula Weight 147.2
Grade ACS/EP/Reagent
Packaging Plastic Bottle
Density 1.85g/cm3 (2°C)
Chemical Name or Material Calcium Chloride, Dihydrate, EMSURE™, For Analysis
Recommended Storage Store without Restrictions