missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride dihydrate, 99%, for biochemistry
CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol
$147.08 - $147.08
Chemical Identifiers
| CAS | 10035-04-8 |
|---|---|
| Molecular Formula | CaCl2·2H2O |
| Molecular Weight (g/mol) | 147.02 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC447325000
|
Thermo Scientific Chemicals
447325000 |
500 g | Plastic Bottle |
Each for $147.08
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10035-04-8 | |
| CaCl2·2H2O | |
| 147.02 |
Specifications
| 175.0°C | |
| 500 g | |
| CaCl2·2H2O | |
| 15,1661 | |
| 147.02 | |
| 99% | |
| Plastic Bottle | |
| 0.8300g/mL |
| 10035-04-8 | |
| 98.5% min. (Complexometry) | |
| CaCl2·2H2O | |
| Clear colorless | |
| 147.02 | |
| Biochemical | |
| 0.83 | |
| Calcium chloride dihydrate |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only