missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, 99%, for biochemistry

Catalog No. AC447325000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447325000 500 g Plastic Bottle
AC447320025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447325000 Supplier Thermo Scientific Chemicals Supplier No. 447325000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O
Molecular Weight (g/mol) 147.02

Specifications

Melting Point 175.0°C
Quantity 500 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula CaCl2·2H2O
Merck Index 15,1661
Solubility Information Clear colorless
Formula Weight 147.02
Percent Purity 99%
Grade Biochemical
Packaging Plastic Bottle
Specific Gravity 0.83
Density 0.8300g/mL
Chemical Name or Material Calcium chloride dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only