missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, 99+%, for analysis

Catalog No. p-4509901
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207780010 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC207780010 Supplier Thermo Scientific Chemicals Supplier No. 207780010
Only null left

CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O
Molecular Weight (g/mol) 147.02

Specifications

Melting Point 175.0°C
Color White
pH 4.5 to 6.5 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 1 kg
Assay Percent Range 99% min. (Complexometry)
Linear Formula CaCl2·2H2O
Fieser 01,104
Merck Index 15,1661
Solubility Information Solubility in water: 1000g/L (0°C). Other solubilities: freely soluble in alcohol
Formula Weight 147.02
Percent Purity ≥99%
Grade Analytical
Packaging Plastic Bottle
Specific Gravity 0.83
Density 0.8300g/mL
Chemical Name or Material Calcium chloride dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only