missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbonate, Purified, Reagents

Catalog No. 01337343
Change view
Click to view available options
:
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01337343 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337343 Supplier Labchem Inc Supplier No. C2161500500A5
Only null left

Calcium Carbonate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym Limestone, Marble, Chalk, Aragonite, Calcite
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.71
Chemical Name or Material Calcium Carbonate, Purified