missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbonate (Powder/USP/FCC), Fisher Chemical™

Catalog No. p-31472
Change view
Click to view available options
Quantity:
3 kg
10 kg
Packaging:
Fiber Drum
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C633 3 kg Poly Pail
C6310 10 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C633 Supplier Thermo Fisher Scientific Supplier No. FLC633
Only null left

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825°C
Color White
pH 8 to 9
Physical Form Solid
Quantity 3 kg
Formula Weight 100.09
Percent Purity 98.0 to 100.5% (FCC)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (FCC/USP), Pass Test (USP)
Loss on Drying 2.0% max. (FCC), 2.0% max. (FCC/USP), 2.0% max. (USP)
Packaging Poly Pail
Chemical Name or Material Calcium Carbonate
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes and skin. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:0
Instability:0