missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Carbonate (Powder/ACS Grade), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLC643
Specifications
| Calcium Carbonate | |
| White | |
| 8 to 9 | |
| 3 kg | |
| CCaO3 | |
| calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
| [Ca++].[O-]C([O-])=O | |
| 100.09 | |
| CHEBI:3311 | |
| ≥99.0% | |
| Pass Test |
| 825°C | |
| 471-34-1 | |
| Solid | |
| ≥99.0 % | |
| MFCD00010906 | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| calcium carbonate | |
| 10112 | |
| 100.09 | |
| Certified ACS | |
| Poly Pail |
Chemical Identifiers
| 471-34-1 | |
| 100.09 | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| 10112 | |
| calcium carbonate |
| CCaO3 | |
| MFCD00010906 | |
| calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
| CHEBI:3311 | |
| [Ca++].[O-]C([O-])=O |
Safety and Handling