missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbonate, Light Powder, U.S.P. - F.C.C., J.T. Baker™

Catalog No. p-5390717
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002512 500 g Poly Bottle
02002511 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002512 Supplier Avantor Supplier No. 130101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Physical Form Powder
Quantity 500 g
IUPAC Name calcium;carbonate
Formula Weight 100.09
Grade FCC/USP
Packaging Poly Bottle
Chemical Name or Material Calcium Carbonate