missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, chelometric standard, ACS, 99.95-100.05%

Catalog No. AA3639936
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3639936 500 g
AA3639922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3639936 Supplier Thermo Scientific Chemicals Supplier No. 03639936

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Calcium carbonate, CCaO3, CAS Number-471-34-1, aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble, 500g, White powder, 99.95-100.05%, CHEBI:3311, 2.93, 207-439-9, 100.09, ACS, VTYYLEPIZMXCLO-UHFFFAOYSA-L, calcium;carbonate | CAS: 471-34-1 | CCaO3 | 100.09 g/mol

Calcium carbonate is a dietary supplement used when the quantity of calcium taken in the diet is not enough. Used as an antacid to relieve heartburn, acid indigestion and upset belly. Practiced in the expression industry. Also widely used as a filler in plastics and is used in metallurgy of iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 800°C (decomposition)
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.95-100.05%
Solubility Information Insoluble in water but soluble in acids.
IUPAC Name calcium;carbonate
Formula Weight 100.09
Grade ACS
Refractive Index 1.6583
Appearance White powder
Density 2.93
Chemical Name or Material Calcium carbonate, For analysis ACS
Show More Show Less
EINECSNumber 207-439-9
RTECSNumber FF9335000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only