missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium carbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E170, precipitated, 98.5-100.5% (based on anhydrous substance), Honeywell Fluka™
Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
$199.26 - $776.87
Chemical Identifiers
| CAS | 471-34-1 |
|---|---|
| Molecular Formula | CCaO3 |
| Molecular Weight (g/mol) | 100.09 |
| MDL Number | MFCD00010906 |
| InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
| Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
| PubChem CID | 10112 |
| ChEBI | CHEBI:3311 |
| SMILES | [Ca++].[O-]C([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 471-34-1 | |
| 100.09 | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| 10112 | |
| [Ca++].[O-]C([O-])=O |
| CCaO3 | |
| MFCD00010906 | |
| calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
| CHEBI:3311 |
Specifications
| 471-34-1 | |
| CCaO3 | |
| MFCD00010906 | |
| 8008338 | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| calcium;carbonate | |
| 10112 | |
| 100.09g/mol | |
| puriss. | |
| Calcium Carbonate(Caco3), Meets analytical specification of Ph. Eur.BP, USP, FCC |
| 1 kg | |
| CaCO3 | |
| NONH for all modes of transport | |
| calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
| [Ca++].[O-]C([O-])=O | |
| 100.09 | |
| CHEBI:3311 | |
| 98.5 to 100.5% (based on anhydrous substance) | |
| 2.93g/mL at 25°C |
Safety and Handling
EINECSNumber : 207-439-9
RTECSNumber : FF9335000