missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, ACS reagent, chelometric standard

Catalog No. p-7077262
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC403801000 100 g
AC403805000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC403801000 Supplier Thermo Scientific Chemicals Supplier No. 403801000
Only null left

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825.0°C
Quantity 100 g
Linear Formula CaCO3
Fieser 01,103; 02,57; 04,67; 05,89
Merck Index 15,1659
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted acids
IUPAC Name calcium carbonate
Formula Weight 100.09
Percent Purity 99.95 to 100.05% (on dried substance) (Complexometry)
Grade ACS Reagent
Chemical Name or Material Calcium carbonate
Show More Show Less
EINECSNumber 207-439-9

RUO – Research Use Only