missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, 99%, extra pure

Catalog No. p-7077348
Change view
Click to view available options
Quantity:
1 kg
25 kg
5 kg
Catalog No. Quantity
AC450680010 1 kg
AC450680050 5 kg
AC450680250 25 kg
3 options

Catalog No. AC450680010

Supplier: Thermo Scientific Chemicals 450680010

Only null left

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825.0°C
Quantity 1 kg
Assay Percent Range 98.5 to 101.5% (Complexometry)
Linear Formula CaCO3
Merck Index 15,1659
Solubility Information (in dilute HCl) Clear colorless to light yellow
IUPAC Name calcium carbonate
Formula Weight 100.09
Percent Purity 99%
Grade Extra Pure
Loss on Drying 2% max. (200°C)
Chemical Name or Material Calcium carbonate
Show More Show Less
EINECSNumber 207-439-9

RUO – Research Use Only