missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, 99%, extra pure

Catalog No. AC450680010
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450680010 1 kg
AC450680050 5 kg
AC450680250 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC450680010 Supplier Thermo Scientific Chemicals Supplier No. 450680010

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825.0°C
Quantity 1 kg
Assay Percent Range 98.5 to 101.5% (Complexometry)
Linear Formula CaCO3
Merck Index 15,1659
Solubility Information (in dilute HCl) Clear colorless to light yellow
IUPAC Name calcium carbonate
Formula Weight 100.09
Percent Purity 99%
Grade Extra Pure
Loss on Drying 2% max. (200°C)
Chemical Name or Material Calcium carbonate
Show More Show Less
EINECSNumber 207-439-9

RUO – Research Use Only