missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, 99%, for analysis, precipitated

Catalog No. p-46028
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
500 g
Packaging:
Plastic bottle
Plastic drum
Catalog No. Quantity Packaging
AC219165000 500 g Plastic bottle
AC219160010 1 kg Plastic bottle
AC219160025 2.5 kg Plastic drum
3 options

Catalog No. AC219165000

Supplier: Thermo Scientific Chemicals 219165000

Only null left

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 98.5% min. (on dried substance) (Complexometry)
Linear Formula CaCO3
Fieser 01,103; 02,57; 04,67; 05,89
Merck Index 15, 1659
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted acids
IUPAC Name calcium;carbonate
Formula Weight 100.09
Percent Purity 99%
Grade Analytical
Loss on Drying 0.5% max. (200°C)
Packaging Plastic bottle
Chemical Name or Material Calcium carbonate, 99%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-439-9
RTECSNumber FF9335000
TSCA TSCA

RUO – Research Use Only