missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, 98%

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

$118.99 - $206.53

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1236536
View Documents
Thermo Scientific Chemicals
01236536
500 g N/A
Only null left
 
AA12365A3
View Documents
Thermo Scientific Chemicals
012365A3
2 kg N/A
Only null left
 
Description

Description

Manufacture of paint, rubber, plastics, etc. In food, cosmetics, and pharmaceuticals. As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizerIt is used medicinally as a calcium supplement or as an antacid. It is also used for industrial purposes. Calcium carbonate is the most widely used mineral in paper, plastics, paints and coatings industries. Hierarchical hollow calcium carbonate particles shows an useful application as anticancer drug carrier. It plays an essential role to catalyze alcoholysis of fats and oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Manufacture of paint, rubber, plastics, etc
  • In food, cosmetics, and pharmaceuticals
  • As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizer
Specifications

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium carbonate
CCaO3
MFCD00010906
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
CHEBI:3311
[Ca++].[O-]C([O-])=O

Specifications

800°C (decomposition)
White
500 g
CCaO3
14,1657
Practically insoluble in water; Soluble in diluted acids
[Ca++].[O-]C([O-])=O
100.09
CHEBI:3311
98%
1.6583
Calcium carbonate
471-34-1
Powder
98%
MFCD00010906
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
10112
100.09
Odorless
2.930 g/mL
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-439-9

RTECSNumber : FF9335000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only