missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, 98+%, pure, heavy powder

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

$159.84 - $159.84

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124670010
SDS View Documents
Thermo Scientific Chemicals
124670010
1 kg Plastic bottle
Each for $159.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
[Ca++].[O-]C([O-])=O
CCaO3
MFCD00010906
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
CHEBI:3311

Specifications

825.0°C
White
1 kg
CCaO3
MFCD00010906
15,1659
Solubility in water: insoluble. Other solubilities: soluble in diluted acids
[Ca++].[O-]C([O-])=O
100.09
CHEBI:3311
≥98%
Plastic bottle
471-34-1
Powder
98% min. (Complexometry)
CaCO3
01,103; 02,57; 04,67; 05,89
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
10112
100.09
Pure
Calcium carbonate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-439-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only