missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbide, Lump, Spectrum™ Chemical

$362.29 - $4378.35

Chemical Identifiers

CAS 75-20-7
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609439
View Documents
Spectrum Chemical Mfg Cor
C1068500GM
500 g Amber Glass Bottle
Each for $362.29
Only null left
 
18609437
View Documents
Spectrum Chemical Mfg Cor
C106825KG
2.5 kg Amber Glass Bottle
Each for $843.47
Only null left
 
18609438
View Documents
Spectrum Chemical Mfg Cor
C106845KGBL
45 kg Fiber Drum
Each for $4,378.35
Only null left
 
Description

Description

Specifications

Chemical Identifiers

75-20-7

Specifications

75-20-7
500 g
Amber Glass Bottle
1
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.