missing translation for 'onlineSavingsMsg'
Learn More

Calcium boride, 99+% (metals basis)

Catalog No. AA4155114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4155114 25 g
AA4155122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4155114 Supplier Thermo Scientific Chemicals Supplier No. 04155114

CAS: 12007-99-7 | B6Ca | 104.938 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12007-99-7
Molecular Formula B6Ca
Molecular Weight (g/mol) 104.938
MDL Number MFCD00148904
InChI Key AWVIDOFQNLOVRC-UHFFFAOYSA-N
Synonym calcium boride, calcium hexaboride, b6.ca, calcium hexaboride,-200 mesh, calcium pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide, calcium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide
PubChem CID 16212529
SMILES B12B3[B-]14B5[B-]23B45.[Ca+2]

Specifications

Melting Point 2235°C
Physical Form Powder
Quantity 25 g
Formula Weight 104.95
Percent Purity ≥99%
Density 2.30 g/mL
Assay (metals basis)
Chemical Name or Material Calcium boride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-525-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only