missing translation for 'onlineSavingsMsg'
Learn More

Calcium boride, 95+%

CAS: 12007-99-7 | B6Ca | 104.938 g/mol

Supplier:  Thermo Scientific Chemicals 04172130

 View more versions of this product

Catalog No. AA4172130

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium boride
12007-99-7
250 g
MFCD00148904
Insoluble in water. Dissolves slowly in acids
B12B3[B-]14B5[B-]23B45.[Ca+2]
16212529
≥95%
2235°C
Powder
B6Ca
calcium boride, calcium hexaboride, b6.ca, calcium hexaboride,-200 mesh, calcium pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide, calcium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide
AWVIDOFQNLOVRC-UHFFFAOYSA-N
104.938
104.95
2.30 g/mL

Chemical Identifiers

12007-99-7
104.938
AWVIDOFQNLOVRC-UHFFFAOYSA-N
16212529
B6Ca
MFCD00148904
calcium boride, calcium hexaboride, b6.ca, calcium hexaboride,-200 mesh, calcium pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide, calcium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide
B12B3[B-]14B5[B-]23B45.[Ca+2]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 234-525-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only