missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Bis(fluorosulfonyl)imide 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| UN Number | 1759 |
| Formula Weight | 400.32 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Calcium Bis(fluorosulfonyl)imide |
Safety and Handling
| TSCA | No |