missing translation for 'onlineSavingsMsg'
Learn More

Pointe Scientific Calcium (Arsenazo III) Reagents

Catalog No. 23666170
Encompass
Change view
Click to view available options
Quantity:
1 x 120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23666170 1 x 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23666170 Supplier Pointe Scientific Supplier No. C7529120

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Calcium reagent: Arsenazo III≥0.15mM, 8-Hydroxyquinoline Sulfonate 5.0mM, buffer, surfactant

Requires

Requires Calibrator/Standard

Quantify calcium levels in serum or heparinized plasma

  • Method: Arsenazo III, Endpoint
  • Format: Liquid (ready to use)
  • Wavelength: 650nm
  • Linearity: 0 to 15mg/dL
  • Expected Values: 8.5 to 10.4mg/dL

For the quantitative determination of Calcium in serum or heparinized plasma.

Specifications

Certifications/Compliance CE Marked
Type Calcium Arsenazo III Reagent Kit
Formulation Calcium reagent: Arsenazo III <0.15mM, 8-Hydroxyquinoline Sulfonate 5mM, Buffer, Surfactant
For Use With (Application) Quantitative determination of Calcium in Serum or Heparinized Plasma
Quantity 1 x 120 mL
Wavelength 650 nm
Form Liquid