missing translation for 'onlineSavingsMsg'
Learn More

Calcium aluminum oxide, 99% (metals basis)

CAS: 12042-68-1 | Al2CaO4 | 158.037 g/mol

$109.97 - $882.66

Chemical Identifiers

CAS 12042-68-1
Molecular Formula Al2CaO4
Molecular Weight (g/mol) 158.037
MDL Number MFCD00049722
InChI Key XFWJKVMFIVXPKK-UHFFFAOYSA-N
Synonym calcium aluminate, calcium aluminum oxide, calcium dialumanoylolate, dialuminium calcium tetraoxide, calcium dialuminate, aluminum calcium oxide, 2alo2.ca, calcium oxido oxo alumane, calcium 2+ dialumanoylolate, calcium aluminate 5g
PubChem CID 159415
IUPAC Name calcium;oxido(oxo)alumane
SMILES [O-][Al]=O.[O-][Al]=O.[Ca+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA2264606
View Documents
Thermo Scientific Chemicals
02264606
5 g
Each for $109.97
Only null left
 
AA2264614
View Documents
Thermo Scientific Chemicals
02264614
25 g
Each for $298.34
Only null left
 
AA2264622
View Documents
Thermo Scientific Chemicals
02264622
100 g
Each for $882.66
Only null left
 
Description

Description

Calcium aluminum oxide is used as a precursor in the production of naphtha steam reforming catalysts, refinery gas steam reforming catalysts, aluminum chlorohydrate, secondary hydrocarbon steam reforming catalysts. It is also used in the production of refractory and cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12042-68-1
158.037
XFWJKVMFIVXPKK-UHFFFAOYSA-N
159415
[O-][Al]=O.[O-][Al]=O.[Ca+2]
Al2CaO4
MFCD00049722
calcium aluminate, calcium aluminum oxide, calcium dialumanoylolate, dialuminium calcium tetraoxide, calcium dialuminate, aluminum calcium oxide, 2alo2.ca, calcium oxido oxo alumane, calcium 2+ dialumanoylolate, calcium aluminate 5g
calcium;oxido(oxo)alumane

Specifications

1600°C
12042-68-1
5 g
Al2CaO4
calcium aluminate, calcium aluminum oxide, calcium dialumanoylolate, dialuminium calcium tetraoxide, calcium dialuminate, aluminum calcium oxide, 2alo2.ca, calcium oxido oxo alumane, calcium 2+ dialumanoylolate, calcium aluminate 5g
XFWJKVMFIVXPKK-UHFFFAOYSA-N
calcium;oxido(oxo)alumane
159415
Odorless
2.981
White
Powder
99% (Metals basis)
MFCD00049722
Insoluble in water.
[O-][Al]=O.[O-][Al]=O.[Ca+2]
158.037
158.02
Hygroscopic
Calcium aluminum oxide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-931-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only