missing translation for 'onlineSavingsMsg'
Learn More

Calcium aluminum oxide, 99% (metals basis)

Catalog No. AA2264622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2264622 100 g
AA2264606 5 g
AA2264614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA2264622 Supplier Thermo Scientific Chemicals Supplier No. 02264622
Only null left

CAS: 12042-68-1 | Al2CaO4 | 158.037 g/mol

Calcium aluminum oxide is used as a precursor in the production of naphtha steam reforming catalysts, refinery gas steam reforming catalysts, aluminum chlorohydrate, secondary hydrocarbon steam reforming catalysts. It is also used in the production of refractory and cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12042-68-1
Molecular Formula Al2CaO4
Molecular Weight (g/mol) 158.037
MDL Number MFCD00049722
InChI Key XFWJKVMFIVXPKK-UHFFFAOYSA-N
Synonym calcium aluminate, calcium aluminum oxide, calcium dialumanoylolate, dialuminium calcium tetraoxide, calcium dialuminate, aluminum calcium oxide, 2alo2.ca, calcium oxido oxo alumane, calcium 2+ dialumanoylolate, calcium aluminate 5g
PubChem CID 159415
IUPAC Name calcium;oxido(oxo)alumane
SMILES [O-][Al]=O.[O-][Al]=O.[Ca+2]

Specifications

Melting Point 1600°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 158.02
Odor Odorless
Sensitivity Hygroscopic
Density 2.981
Chemical Name or Material Calcium aluminum oxide
Show More Show Less
EINECSNumber 234-931-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only