missing translation for 'onlineSavingsMsg'
Learn More

Puritan™ Calcium Alginate Swabs

Calcium alginate applicators permit superior culture isolation from areas such as the eyes, ears, nasopharynx, urethra and pediatric ENT

Supplier:  Puritan™ 258062WA

 View more versions of this product

Catalog No. 14959105



Only null left

Description

Description

Calcium alginate applicators permit superior culture isolation from areas such as the eyes, ears, nasopharynx, urethra and pediatric ENT

  • Processing purges any detectable bactericidal agent in the calcium alginate wool tips, eliminating the need for charcoal as a component for transport systems
  • Calcium alginate fibers lack fatty acids that inhibit many gonorrhea strains
  • Calcium alginate fibers are slowly dissolved by body fluids and tissues, eliminating the potential for reactions to foreign matter

Specifications

Chemical Identifiers

CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
Swab
Sterile
Calcium Alginate Fiber

Specifications

CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
6 in.
Swab
15.2 cm
14.7 cm
1.6 cm
0.21 in.
Wood
0.10 in.
Specimen Collection/Wound Care
2.5 mm
5.81 in.
0.641 in.
0.55 cm
Sterile
Calcium Alginate Fiber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions